Learn More
Ace Glass, Inc. 25mm ID x 30cm chromatography column, 24/40 outer top, 250mL reservoir, porosity B filter disc
SDP
Catalog No. 50972601
Encompass_Preferred
25mm I.D. x 30cm chromatography column with 250mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a 24/40 outer joint. Includes porosity B filter disc.
Catalog No. 50-972-601 Supplier Ace Glass, Inc. Supplier No. 5907141
25mm I.D. x 30cm chromatography column with 250mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a 24/40 outer joint. Includes porosity B filter disc.