Learn More
Chemglass Life Sciences 24/40 3-Port Adapter, Complete with (3) Nuts, (3)Ferrules, and (2) Plugs
SDP
Catalog No. CG103712
Encompass_Preferred
24/40 3-Port Adapter, Complete with (3) Nuts, (3)Ferrules, and (2) Plugs. Replacement 3-port adapter for item CG103710. Comes with 3 nuts, 3 ferrules, and 2 plugs. Complete adapter (CG103710) is supplied with 90¿ valve, 3-port 24/40 PTFE Adapter and 10ft of 1/8¿ OD PTFE tubing.
Catalog No. CG103712 Supplier Chemglass Life Sciences Supplier No. CG103712
24/40 3-Port Adapter, Complete with (3) Nuts, (3)Ferrules, and (2) Plugs. Replacement 3-port adapter for item CG103710. Comes with 3 nuts, 3 ferrules, and 2 plugs. Complete adapter (CG103710) is supplied with 90¿ valve, 3-port 24/40 PTFE Adapter and 10ft of 1/8¿ OD PTFE tubing.