Learn More
Ace Glass, Inc. 24/40 30.5cm distillation column
SDP
Catalog No. NC0672394
Encompass_Preferred
30.5cm Hemple distillation column with 24/40 standard taper joints, and indentations to support packing. Length in centimeters refers to the length from the packing support to the bottom of the outer ground joint.
Catalog No. NC0672394 Supplier Ace Glass, Inc. Supplier No. 657206
30.5cm Hemple distillation column with 24/40 standard taper joints, and indentations to support packing. Length in centimeters refers to the length from the packing support to the bottom of the outer ground joint.