Learn More
Ace Glass, Inc. 24/40 38.1cm distillation column
SDP
Catalog No. 50973275
Encompass_Preferred
Distillation column with 24/40 standard taper joints and perforated glass disc for packing support. 38.1cm length refers to the length from the packing support to the bottom of the outer ground joint.
Catalog No. 50-973-275 Supplier Ace Glass, Inc. Supplier No. 657310
Distillation column with 24/40 standard taper joints and perforated glass disc for packing support. 38.1cm length refers to the length from the packing support to the bottom of the outer ground joint.