Learn More
Ace Glass, Inc. 24/40 50.8cm distillation column
SDP
Catalog No. 50978830
Encompass_Preferred
50.8cm Hemple distillation column with 24/40 standard taper joints, and indentations to support packing. Length in centimeters refers to the length from the packing support to the bottom of the outer ground joint.
Catalog No. 50-978-830 Supplier Ace Glass, Inc. Supplier No. 657212
50.8cm Hemple distillation column with 24/40 standard taper joints, and indentations to support packing. Length in centimeters refers to the length from the packing support to the bottom of the outer ground joint.