Learn More
Ace Glass, Inc. 24/40 50mL distillation receiver
SDP
Catalog No. 50973303
Encompass
50mL all-purpose receiver for general distillation work. Equipped with a complete stopcock manifold to allow contents of the receiver to be removed to a single receiving flask without disturbing the vacuum of the system
Catalog No. 50-973-303 Supplier Ace Glass, Inc. Supplier No. 663704
50mL all-purpose receiver for general distillation work. Equipped with a complete stopcock manifold to allow contents of the receiver to be removed to a single receiving flask without disturbing the vacuum of the system