Learn More
Ace Glass, Inc. 50mm ID x 30cm chromatography column, 24/40 outer top, 500mL reservoir, without filter disc
SDP
Catalog No. 50972598
Encompass_Preferred
50mm I.D. x 30cm chromatography column with 500mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a 24/40 outer joint.
Catalog No. 50-972-598 Supplier Ace Glass, Inc. Supplier No. 5907124
50mm I.D. x 30cm chromatography column with 500mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a 24/40 outer joint.