Learn More
Chemglass Life Sciences Adapter, Hose, Airfree, Schlenk, 24/40 Joint Size, 7.0mm Hose Connection size, Style A
SDP
Catalog No. AF045503
Encompass_Preferred
Adapter, Hose, Airfree, Schlenk, 24/40 Joint Size, 7.0mm Hose Connection size, Style A. Adapters for use with manifolds such as AF-0450 through AF-0454 to permit port connection to standard rubber hose. Adapters have hooks just below the grind for use with CG-110 stainless steel springs. The 24/40 size is available with a constriction above the serrated hose connection to permit bowl area to be filled with glass wool preventing solids from being drawn into the manifold. Size of hose connection is at the middle serration.
Catalog No. AF045503 Supplier Chemglass Life Sciences Supplier No. AF045503
Adapter, Hose, Airfree, Schlenk, 24/40 Joint Size, 7.0mm Hose Connection size, Style A. Adapters for use with manifolds such as AF-0450 through AF-0454 to permit port connection to standard rubber hose. Adapters have hooks just below the grind for use with CG-110 stainless steel springs. The 24/40 size is available with a constriction above the serrated hose connection to permit bowl area to be filled with glass wool preventing solids from being drawn into the manifold. Size of hose connection is at the middle serration.