Learn More
Ace Glass, Inc. 24/40 ADAPT VAC TAKE OFF 24/40 ADAPT VAC TAKE OFF
SDP
Supplier:  Ace Glass, Inc. 519308
Encompass_Preferred
Stem below standard taper joint is 30mm. Hose connection is size C.
Catalog No. 50-955-6965