Learn More
Ace Glass, Inc. Adapter, 75°, 10/30 outer top, 24/40 inner side and bottom
SDP
Catalog No. 50971701
Encompass_Preferred
75 degree angle adapter with 10/30 standard taper outer top joint, and 24/40 standard taper inner side joint and bottom joint.
Catalog No. 50-971-701 Supplier Ace Glass, Inc. Supplier No. 509010
75 degree angle adapter with 10/30 standard taper outer top joint, and 24/40 standard taper inner side joint and bottom joint.