Learn More
Ace Glass, Inc. 24/40 ADAPTER CLAISEN
SDP
Catalog No. 509556937
Encompass_Preferred
Claisen adapter with parallel side arm, 24/40 outer joints at top, and 24/40 inner joint at bottom.
Catalog No. 50-955-6937 Supplier Ace Glass, Inc. Supplier No. 505510
Claisen adapter with parallel side arm, 24/40 outer joints at top, and 24/40 inner joint at bottom.