Learn More
Ace Glass, Inc. Adapter, 24/40, 3/8in size D hose connection, #7 Ace-Thred, complete with 5029-10 nylon bushing and 7855-704 O-ring
SDP
Supplier:  Ace Glass, Inc. 526116
Encompass_Preferred
Adapter with a 24/40 joint at the bottom, 3/8 size D serrated hose connection, and a #7 Ace-Thred at top. Complete with nylon bushing and O-ring.
Catalog No. 50-955-6995