Learn More
Ace Glass, Inc. Adapter, hose connection, bent, 24/40
SDP
Catalog No. 509556974
Encompass_Preferred
Adapter with a 3/8-inch 90-degree hose connection top, and an inner 24/40 standard taper joint bottom.
Catalog No. 50-955-6974 Supplier Ace Glass, Inc. Supplier No. 520510
Adapter with a 3/8-inch 90-degree hose connection top, and an inner 24/40 standard taper joint bottom.