Learn More
Ace Glass, Inc. Adapter, 24/40, 3/8in size D hose connection, #15 Ace-Thred, glass only
SDP
Catalog No. 50978340
Encompass_Preferred
Adapter with a 24/40 joint at the bottom, 3/8 size D serrated hose connection, and a #15 Ace-Thred at top. Does not include nylon bushing or O-ring.
Catalog No. 50-978-340 Supplier Ace Glass, Inc. Supplier No. 526156
Adapter with a 24/40 joint at the bottom, 3/8 size D serrated hose connection, and a #15 Ace-Thred at top. Does not include nylon bushing or O-ring.