Learn More
Chemglass Life Sciences Adapter, Inlet, 24/40 Outer Joint, 2mm Glass Stpk, Airfree, Schlenk
SDP
Catalog No. AF050513
Encompass_Preferred
Adapter, Inlet, 24/40 Outer Joint, 2mm Glass Stpk, Airfree, Schlenk. Gas inlet adapter with a 2mm glass stopcock having a constant pressure retention device. Available with a choice of inner or outer standard taper joint. Top arm is 9mm O.D. medium wall tubing for use with CG-3022 septum. Overall height of the 14/20 size is approx. 95mm while the 24/40 is approx. 120mm.
Catalog No. AF050513 Supplier Chemglass Life Sciences Supplier No. AF050513
Adapter, Inlet, 24/40 Outer Joint, 2mm Glass Stpk, Airfree, Schlenk. Gas inlet adapter with a 2mm glass stopcock having a constant pressure retention device. Available with a choice of inner or outer standard taper joint. Top arm is 9mm O.D. medium wall tubing for use with CG-3022 septum. Overall height of the 14/20 size is approx. 95mm while the 24/40 is approx. 120mm.