Learn More
Ace Glass, Inc. 24/40 ADAPTER OFFSET 24/40 ADAPTER OFFSET
SDP
Catalog No. 509556927
Encompass_Preferred
Adapter, 10degree Offset, Ace Glass, no. 7 Ace-Thred top joint, standard taper 24/40 bottom joint, Accepts thermometers or bleed tubes with up to 7mm dia., Ideal for use with multi-necked flasks, Includes nylon bushing and FETFE O-ring
Catalog No. 50-955-6927 Supplier Ace Glass, Inc. Supplier No. 503222
Adapter, 10degree Offset, Ace Glass, no. 7 Ace-Thred top joint, standard taper 24/40 bottom joint, Accepts thermometers or bleed tubes with up to 7mm dia., Ideal for use with multi-necked flasks, Includes nylon bushing and FETFE O-ring