Learn More
Chemglass Life Sciences Joint Size: 24/40, Total Length: min. 120 (mm), max. 137 (mm), Bellows Adapter
SDP
Catalog No. 50871884
Encompass_Preferred
Adapter is constructed completely of PTFE and is ideal for eliminating stress between standard joints. Can be bent up to 120¿. Top outer and lower inner are of the same size. Joint Size: 24/40, Total Length: min. 120 (mm), max. 137 (mm)
Catalog No. 50-871-884 Supplier Chemglass Life Sciences Supplier No. CG100420
Adapter is constructed completely of PTFE and is ideal for eliminating stress between standard joints. Can be bent up to 120¿. Top outer and lower inner are of the same size. Joint Size: 24/40, Total Length: min. 120 (mm), max. 137 (mm)