Learn More
Chemglass Life Sciences Adapter, Distillation, Claisen, 24/40 Joint Size, 10/30 Thermometer Joint For 75mm Immersion, 198mm H x 200mm W
SDP
Catalog No. CG101501
Encompass_Preferred
Adapter, Distillation, Claisen, 24/40 Joint Size, 10/30 Thermometer Joint For 75mm Immersion, 198mm H x 200mm W. Distilling adapter having an outer member on the vertical side tube for use with a thermometer of the listed joint size and immersion. Sidearm has a standard taper inner drip tip joint while vertical tube has a top outer and lower inner joint, all being the same size. FOR THERMOMETERS SEE CG-3506-N.
Catalog No. CG101501 Supplier Chemglass Life Sciences Supplier No. CG101501
Adapter, Distillation, Claisen, 24/40 Joint Size, 10/30 Thermometer Joint For 75mm Immersion, 198mm H x 200mm W. Distilling adapter having an outer member on the vertical side tube for use with a thermometer of the listed joint size and immersion. Sidearm has a standard taper inner drip tip joint while vertical tube has a top outer and lower inner joint, all being the same size. FOR THERMOMETERS SEE CG-3506-N.