Learn More
Chemglass Life Sciences Distilling Receiver, 10mL, Moisture Test, Merlic, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 290mm Height x 105mm Width
SDP
Catalog No. CG126406
Encompass_Preferred
Distilling Receiver, 10mL, Moisture Test, Merlic, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 290mm Height x 105mm Width. Modified Dean-Stark trap allows for more efficient cooling and separation of the azeotrope during distillation. Graduated from 0 to 10ml in 0.2mL subdivisions.Design suggested by Dr. Craig Merlic, University of California, Los Angeles, California.
Catalog No. CG126406 Supplier Chemglass Life Sciences Supplier No. CG126406
Distilling Receiver, 10mL, Moisture Test, Merlic, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 290mm Height x 105mm Width. Modified Dean-Stark trap allows for more efficient cooling and separation of the azeotrope during distillation. Graduated from 0 to 10ml in 0.2mL subdivisions.Design suggested by Dr. Craig Merlic, University of California, Los Angeles, California.