Learn More
Ace Glass, Inc. Joint clips, 24/40, Keck; type, POM acetal resin, not for positive pressure use, green, pk/10
SDP
Catalog No. 509557892
Encompass
24/40 green Keck type joint clips made from POM acetal resin. Clips will not scratch the glass, and are resistant to concentrated alkalies and dilute acids. Useful temp range: -40 to 140 C. Not for pressure applications. Pk/10.
Catalog No. 50-955-7892 Supplier Ace Glass, Inc. Supplier No. 759824
24/40 green Keck type joint clips made from POM acetal resin. Clips will not scratch the glass, and are resistant to concentrated alkalies and dilute acids. Useful temp range: -40 to 140 C. Not for pressure applications. Pk/10.