Learn More
Ace Glass, Inc. Receiver, distillation, 24/40, PTFE plug
SDP
Catalog No. 50973302
Encompass_Preferred
Especially designed receiver to draw samples during distillation or to return the condensed vapor to the flask. Capacity 125ml, available with 1:5 PTFE plug.
Catalog No. 50-973-302 Supplier Ace Glass, Inc. Supplier No. 663520
Especially designed receiver to draw samples during distillation or to return the condensed vapor to the flask. Capacity 125ml, available with 1:5 PTFE plug.