Learn More
Ace Glass, Inc. Receiver, distillation, 24/40, glass plug
SDP
Catalog No. 50978840
Encompass_Preferred
Especially designed receiver to draw samples during distillation or to return the condensed vapor to the flask. Capacity 125ml.
Catalog No. 50-978-840 Supplier Ace Glass, Inc. Supplier No. 663510
Especially designed receiver to draw samples during distillation or to return the condensed vapor to the flask. Capacity 125ml.