Learn More
Ace Glass, Inc. 24/40 variable reflux ratio distilling head with 10/30 thermometer joint
SDP
Catalog No. 50973289
Encompass
24/40 variable reflux ratio distilling head with 10/30 thermometer joint takes 76mm immersion thermometers. Side arm joint is 24/40. Head is constructed so that satisfactory operation can be maintained under any pressure condition.
Catalog No. 50-973-289 Supplier Ace Glass, Inc. Supplier No. 660410
24/40 variable reflux ratio distilling head with 10/30 thermometer joint takes 76mm immersion thermometers. Side arm joint is 24/40. Head is constructed so that satisfactory operation can be maintained under any pressure condition.