Learn More
Ace Glass, Inc. 24/40 inner joint, stainless steel, 1/2" male NPT
SDP
Catalog No. 50979406
Encompass
24/40 inner joint made of stainless steel. Joint is machined with male NPT thread, manufactured from 18-8 type 303 free machining stainless steel, and supplied in accordance with CS21-58 of the NIST.
Catalog No. 50-979-406 Supplier Ace Glass, Inc. Supplier No. 7651220
24/40 inner joint made of stainless steel. Joint is machined with male NPT thread, manufactured from 18-8 type 303 free machining stainless steel, and supplied in accordance with CS21-58 of the NIST.