Learn More
Ace Glass, Inc. 24/40 JOINTS O-RING SEAL -115
SDP
Catalog No. 50979403
Encompass_Preferred
Standard taper inner member ground joint with groove for O-Ring. For use with outer members 7565, 7566, 7567, 7651, and 7652. Leakproof at high vacuum (up to 1 x 10-8mm Hg.). O-Ring permits use without grease. Supplied with one FETFE O-Ring. For replacement O-Rings, see 7855.
Catalog No. 50-979-403 Supplier Ace Glass, Inc. Supplier No. 764812
Standard taper inner member ground joint with groove for O-Ring. For use with outer members 7565, 7566, 7567, 7651, and 7652. Leakproof at high vacuum (up to 1 x 10-8mm Hg.). O-Ring permits use without grease. Supplied with one FETFE O-Ring. For replacement O-Rings, see 7855.