Learn More
Chemglass Life Sciences 24/40 Outer Joint, Dewar Seal, 55mm OD Flare
SDP
Catalog No. CG13134
Encompass_Preferred
24/40 Outer Joint, Dewar Seal, 55mm OD Flare. Used in construction of apparatus requiring the outer member to be jacketed. Configuration of flare permits seal to be made without having to regrind the joint.
Catalog No. CG13134 Supplier Chemglass Life Sciences Supplier No. CG13134
24/40 Outer Joint, Dewar Seal, 55mm OD Flare. Used in construction of apparatus requiring the outer member to be jacketed. Configuration of flare permits seal to be made without having to regrind the joint.