Learn More
Chemglass Life Sciences 24/40 Lower Joint Adapter Only, 10mm Stir Bearing, Mechanical Seal, 1-3/4in Threaded Connection, Complete with O-Rings
SDP
Supplier:  Chemglass Life Sciences CG2077M37
Encompass_Preferred
• Dry running mechanical seal rotates with the shaft providing for a vacuum and pressure tight seal; maximum vacuum rating is 5 Torr, maximum pressure is 2-3 PSI• Unlike conventional PTFE Bearings, bearing will not “flake” contaminating your reaction• All seals are made using perfluorinated O-rings providing outstanding resistance to the most aggressive chemicals• Bearing seals are rated at up to 2000 RPM* with the seal life being 3000 hoursOur mechanical seal bearing is the only design on the market that will permit you to use your current glass, metal or PTFE covered stir shaft. The design allows you to have the flexibility of vertical adjustment and still choose your agitator diameter and style. Bearing has a ferrule and locking nut to ensure that the seal rotates properly, and is supplied complete with a gold-plated wrench. Lower joint adapters are supplied complete with perfluorinated O-rings and a loosening nut.PLEASE NOTE: Bearing body and lower joint adapter must be
Catalog No. NC0349541