Learn More
Ace Glass, Inc. Adapter, #7 Ace-Thred top, 24/40 inner bottom, 8mm hole nylon bushing with FETFE O-ring
SDP
Catalog No. 509556907
Encompass_Preferred
#7 Ace-Thred mini adapter with 24/40 inner joint bottom, and nylon bushing with FETFE O-ring. Used for probes, thermometers, bleed tubes, and electrodes. Adapter will accomm ODate items up to 8mm diameter. Suitable for vacuum work.
This item is not returnable. View return policy
Catalog No. 50-955-6907 Supplier Ace Glass, Inc. Supplier No. 5028119
#7 Ace-Thred mini adapter with 24/40 inner joint bottom, and nylon bushing with FETFE O-ring. Used for probes, thermometers, bleed tubes, and electrodes. Adapter will accomm ODate items up to 8mm diameter. Suitable for vacuum work.