Learn More
Ace Glass, Inc. Adapter, #7 Ace-Thred top, 24/40 inner bottom, nylon bushing with FETFE O-ring
SDP
Catalog No. 509556902
Encompass_Preferred
#7 Ace-Thred mini adapter with 19/22 inner joint bottom, and nylon bushing with FETFE O-ring. Used for probes, thermometers, bleed tubes, and electrodes. Adapter will accomm ODate items up to 7mm diameter. Suitable for vacuum work.
Catalog No. 50-955-6902 Supplier Ace Glass, Inc. Supplier No. 502830
#7 Ace-Thred mini adapter with 19/22 inner joint bottom, and nylon bushing with FETFE O-ring. Used for probes, thermometers, bleed tubes, and electrodes. Adapter will accomm ODate items up to 7mm diameter. Suitable for vacuum work.