Learn More
Ace Glass, Inc. Adapter, #7 Ace-Thred top, 24/40 inner bottom, nylon bushing with PTFE ferrule
SDP
Catalog No. 509556903
Encompass_Preferred
#7 Ace-Thred mini adapter with 24/40 inner joint bottom, and nylon bushing with PTFE ferrule. Used for probes, thermometers, bleed tubes, and electrodes. Adapter will accomm ODate items up to 7mm diameter. Suitable for vacuum work.
Catalog No. 50-955-6903 Supplier Ace Glass, Inc. Supplier No. 502831
#7 Ace-Thred mini adapter with 24/40 inner joint bottom, and nylon bushing with PTFE ferrule. Used for probes, thermometers, bleed tubes, and electrodes. Adapter will accomm ODate items up to 7mm diameter. Suitable for vacuum work.