Learn More
Ace Glass, Inc. Trap, vacuum, complete, bent vertical tube, 24/40, 28mm outer body OD, 200mm outer body length below joint, 10mm inner & side tube OD
SDP
Catalog No. 509558572
Encompass_Preferred
Trap, vacuum, complete, bent vertical tube, 24/40, 28mm outer body OD, 200mm outer body length below joint, 10mm inner & side tube OD
Catalog No. 50-955-8572 Supplier Ace Glass, Inc. Supplier No. 875337
Trap, vacuum, complete, bent vertical tube, 24/40, 28mm outer body OD, 200mm outer body length below joint, 10mm inner & side tube OD