Learn More
Chemglass Life Sciences 24/40 Outer Joint, Full Length, on 22mm OD x 1.8mm W tubing
SDP
Catalog No. CG10206
Encompass_Preferred
Standard taper outer joint tooled on the same size tubing as the CG-100 inner joints. All CG-102 outer joints are supplied standard with rings, but are available tooled without rings on a special order basis. 24/40 Outer Joint, Full Length, on 22mm OD x 1.8mm W tubing.
Catalog No. CG10206 Supplier Chemglass Life Sciences Supplier No. CG10206
Standard taper outer joint tooled on the same size tubing as the CG-100 inner joints. All CG-102 outer joints are supplied standard with rings, but are available tooled without rings on a special order basis. 24/40 Outer Joint, Full Length, on 22mm OD x 1.8mm W tubing.