Learn More
Chemglass Life Sciences 24/40 Outer Joint, 28mm X 2.0W Tubing
SDP
Catalog No. 501531891
Encompass_Preferred
24/40 Outer Joint, 28mm X 2.0W Tubing. Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint.
Catalog No. 50-153-1891 Supplier Chemglass Life Sciences Supplier No. CGB10106
24/40 Outer Joint, 28mm X 2.0W Tubing. Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint.