Learn More
Ace Glass, Inc. 24/40 outer joint, stainless steel, 1/2" male NPT
SDP
Catalog No. 50975255
Encompass
24/40 outer joint made of stainless steel. Joint is machined with male NPT thread, manufactured from 18-8 type 303 free machining stainless steel, and supplied in accordance with CS21-58 of the NIST.
Catalog No. 50-975-255 Supplier Ace Glass, Inc. Supplier No. 7651222
24/40 outer joint made of stainless steel. Joint is machined with male NPT thread, manufactured from 18-8 type 303 free machining stainless steel, and supplied in accordance with CS21-58 of the NIST.