Learn More
Ace Glass, Inc. 24/40 outer joint, stainless steel, unthreaded
SDP
Catalog No. 50975252
Encompass
24/40 outer joint made of stainless steel. Joint is unthreaded and machined from 18-8 type 303 free machining stainless steel and supplied in accordance with CS21-58 of the NIST.
Catalog No. 50-975-252 Supplier Ace Glass, Inc. Supplier No. 765122
24/40 outer joint made of stainless steel. Joint is unthreaded and machined from 18-8 type 303 free machining stainless steel and supplied in accordance with CS21-58 of the NIST.