Learn More
Ace Glass, Inc. 24/40 PTFE SLEEVE 12/PK 24/40 PTFE SLEEVE 12/PK
SDP
Catalog No. 509557937
Encompass
PTFE SLEEVES, provides a vacuum-tight seal to a ground joint without grease wherever used. The sleeve is an elongated cone of polytetrafluorethylene film, 0.076 mm (.003 in. ) thick, to fit over the male ground glass cone
Catalog No. 50-955-7937 Supplier Ace Glass, Inc. Supplier No. 7643108
PTFE SLEEVES, provides a vacuum-tight seal to a ground joint without grease wherever used. The sleeve is an elongated cone of polytetrafluorethylene film, 0.076 mm (.003 in. ) thick, to fit over the male ground glass cone