Learn More
Ace Glass, Inc. 24/40 PTFE SLEEVE 3/PK 24/40 PTFE SLEEVE 3/PK
SDP
Supplier:  Ace Glass, Inc. 764108
Encompass
PTFE sleeves with 24/40 joint size and .4mm wall thickness. Sturdy and reusable, features reinforced gripping ring for easy and safe removal from standard taper ground joints. Pack of 3.
Catalog No. 50-955-7921