Learn More
Ace Glass, Inc. 24/40 PTFE Stopper with Polypropylene Extraction Nut, Solid
SDP
Catalog No. 50977609
Encompass
24/40 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.
Catalog No. 50-977-609 Supplier Ace Glass, Inc. Supplier No. 1263618
24/40 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.