Learn More
Chemglass Life Sciences Stirrer Bearing, PTFE, Chem-Stir, Complete, 10mm, 24/40 Inner Joint
SDP
Catalog No. 50873397
Encompass_Preferred
Stirrer Bearing, PTFE, Chem-Stir, Complete, 10mm, 24/40 Inner Joint. Grease-Free PTFE stirrer bearing designed for use under moderate vacuum applications. The bearing consists of a threaded glass adapter having an inner joint, a set of PTFE washer style bearings, a stainless steel spring, nylon alignment ring and Celcon compression cap. The unique design of this bearing provides hours of continuous vacuum operation, with a minimum of "flaking" of the PTFE bearing surfaces.PLEASE NOTE: FOR USE ONLY WITH CG-2078 OR CG-2079 PRECISION GROUND AND POLISHED STIRRER SHAFTS.
Catalog No. 50-873-397 Supplier Chemglass Life Sciences Supplier No. CG207601
Stirrer Bearing, PTFE, Chem-Stir, Complete, 10mm, 24/40 Inner Joint. Grease-Free PTFE stirrer bearing designed for use under moderate vacuum applications. The bearing consists of a threaded glass adapter having an inner joint, a set of PTFE washer style bearings, a stainless steel spring, nylon alignment ring and Celcon compression cap. The unique design of this bearing provides hours of continuous vacuum operation, with a minimum of "flaking" of the PTFE bearing surfaces.PLEASE NOTE: FOR USE ONLY WITH CG-2078 OR CG-2079 PRECISION GROUND AND POLISHED STIRRER SHAFTS.