Learn More
Ace Glass, Inc. 24/40 inner to 15mm Ace-Thred maxi adapter, glass only
SDP
Catalog No. 509558152
Encompass_Preferred
24/40 inner bearing to 15mm Ace-Thred Maxi Adapter, glass only. Part of the Self-Aligning, Trubore Three-Piece Bearing.
Catalog No. 50-955-8152 Supplier Ace Glass, Inc. Supplier No. 804215
24/40 inner bearing to 15mm Ace-Thred Maxi Adapter, glass only. Part of the Self-Aligning, Trubore Three-Piece Bearing.