Learn More
WELCH MATERIALS INC DoprahTM 24-400 Black Closed Top PP Cap with White PE Septa. 100pcs/pk, 10pk/cs.
Catalog No. NC2078224
Encompass_Preferred
24-400 Black Closed Top PP Cap with White PE Septa. 100pcs/pk, 10pk/cs.
Catalog No. NC2078224 Supplier WELCH MATERIALS INC Supplier No. DI121710
24-400 Black Closed Top PP Cap with White PE Septa. 100pcs/pk, 10pk/cs.