Learn More
Chemglass Life Sciences 24-410 G.P.I. Glass Thread, 22mm OD Tubing, 150mm OAL
SDP
Catalog No. CG19008
Encompass_Preferred
24-410 G.P.I. Glass Thread, 22mm OD Tubing, 150mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread.PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.
Catalog No. CG19008 Supplier Chemglass Life Sciences Supplier No. CG19008
24-410 G.P.I. Glass Thread, 22mm OD Tubing, 150mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread.PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.