Learn More
Hach Company 24 Btl 1L Glass SD900 Port.
Supplier:  Hach Company PS241000
Encompass
Kit-24 Bottle-1L PE-for SD900 Portable Standard SamplerStandard Portable Bottle Kit with (24) 1 L poly bottles with caps retainer and full bottle shut off Bottle Tray for 8/24 Bottle Sampling needs to be purchased separately
Catalog No. 50-002-07313