Learn More
Hach Company 24 Btl 350mL Glass SD900 Port
Supplier:  Hach Company PS240350
Encompass
Kit-24 Bottle-350mL-Glass-for SD900 Portable Standard SamplerStandard Portable Bottle Kit with (24) 350 mL glass bottles with caps retainer and full bottle shut off
Catalog No. 50-002-07112