Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ 3500xL Genetic Analyzer 24-Capillary Array, 36 cm
GSA_VA

Catalog No. 4404687
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Array
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-046-87 1 Array
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-046-87 Supplier Applied Biosystems™ Supplier No. 4404687
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use for fragment analysis and rapid sequencing applications.

The internally uncoated capillaries are supplied in preassembled sets of 24 capillaries per array, with a built-in frame for easy installation.

Order Info

Each

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capillary Length 36 cm
Content And Storage Store this array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to your genetic analyser's maintenance, troubleshooting and reference guide.
Description 24-Capillary Array, 36cm Capillary, Internally Uncoated Capillaries, Human Identification Applications, For Use With 3500xL Genetic Analyzer, Room Temperature
For Use With (Equipment) 3500xL Genetic Analyzer
For Use With (Application) Sequencing, Human Identification
Product Type 3500xL Genetic Analyzer
Quantity 1 Array
Shipping Condition Room Temperature
My array is stuck in the pump on my Applied Biosystems 3500/3500xL Genetic Analyzer, even after loosening the knob. What can I do?

If the array is stuck in the pump, even after loosening the knob, squirt around the edge of the array head with distilled water and try to remove it again. If the array will not move, a service call may be required. Do not use excessive force or tools to try and remove it.

When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?

Some of the other causes of an Unstable Electrophoresis current detected error message are:

1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues

Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.

I have an Applied Biosystems 3500/3500xL Genetic Analyzer. When do my consumables expire?

If we do not specify the expiry date or number of uses, the warranty will last for 12 months from the date of shipment.

How often should I change the array on my Applied Biosystems 3500/3500xL Genetic Analyzer?

The Applied Biosystems 3500/3500xl Genetic Analyzer capillary arrays are warrantied for a specific number of runs or until the expiration date printed on the array package, whichever comes first. If you notice that you are starting to get reduced data quality (e.g., shorter reads, trailing peaks in all 4 colors) even with the Sequencing Standard, these may be signs that the array requires changing.

My Applied Biosystems 3500/3500xL Genetic Analyzer consumables have expired, but how long past expiration can I really use them?

Thermo Fisher Scientific can only provide warranty on performance on non-expired reagents. Using reagents past their expiration date is a risk and may compromise your data integrity.

Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?

If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.