Learn More
V&P Scientific Inc ASPIRATION WAND, 24 Channel, 384 Well, Standard, 21 Gauge, SS Tubes, 11mm Long, 9mm Spacing, Polypropylene
Catalog No. NC0172214
Encompass
ASPIRATION WAND, 24 Channel, 384 Well, Standard Plate, 21 Gauge, Stainless Steel Tubes, 11mm Long, 9mm Center Spacing, Polypropylene, Multi-spense Locating Pins, and Female Luer Lok
Catalog No. NC0172214 Supplier V&P Scientific Inc Supplier No. VP 186LP
ASPIRATION WAND, 24 Channel, 384 Well, Standard Plate, 21 Gauge, Stainless Steel Tubes, 11mm Long, 9mm Center Spacing, Polypropylene, Multi-spense Locating Pins, and Female Luer Lok