Learn More
Integra Biosciences Corp 24 CHANNEL PIPETE HEAD 50-1250
SDP
Catalog No. NC2946116
Encompass
  • Enhances liquid handling efficiency and reproducibility in laboratories.
  • Facilitates rapid and precise transfers for various applications.
  • Compatible with VIAFLO 96 and VIAFLO 384 electronic pipettes.
  • Interchangeable design allows switch between channel configurations.
  • Designed for workflows in cell biology, reagent addition, and media transfers.
  • GripTip system ensures secure tip attachment and prevents leaks.
  • High throughput fills 24-well plates quickly.
  • Minimizes human error and enhances accuracy in pipetting.
  • Allows integration into existing VIAFLO systems in under a minute.
  • Pipetting head format for electronic multichannel pipette.
Catalog No. NC2946116 Supplier Integra Biosciences Corp Supplier No. 6124
Add to Cart
Add to Cart
Request a Quote
  • Enhances liquid handling efficiency and reproducibility in laboratories.
  • Facilitates rapid and precise transfers for various applications.
  • Compatible with VIAFLO 96 and VIAFLO 384 electronic pipettes.
  • Interchangeable design allows switch between channel configurations.
  • Designed for workflows in cell biology, reagent addition, and media transfers.
  • GripTip system ensures secure tip attachment and prevents leaks.
  • High throughput fills 24-well plates quickly.
  • Minimizes human error and enhances accuracy in pipetting.
  • Allows integration into existing VIAFLO systems in under a minute.
  • Pipetting head format for electronic multichannel pipette.