Learn More
Electron Microscopy Sciences 24 Color Code Spin Bar Assortment Box
SDP
Supplier:  Electron Microscopy Sciences 6451024
Encompass_Preferred
24 color coded octagon magnetic stirring bars: PTFE with pivot ring, consists of 1 each: Yellow, Red And Blue: 1/2"x1/8", 5/8"x 5/16", 7/8"x 5/16", 1/2"x 5/16", 1"x 5/16", 1.5"x 5/16", 2"x 5/16", 3"x1/2".
Catalog No. 50-270-20