Learn More
AccuStandard, Inc 24 COMP VOC MIX IN METH 5X1ML
SDP
Catalog No. NC9769945
Encompass
Additions to Method 524.2. 100 mL. 1000 ?g/mL in 2-5% Nitric Acid.
Catalog No. NC9769945 Supplier AccuStandard, Inc Supplier No. M524RBPAK
Additions to Method 524.2. 100 mL. 1000 ?g/mL in 2-5% Nitric Acid.