Learn More
Analytical Sales & Services 24 Deep Well Plate, V-Bottom
Catalog No. 5000804610
Encompass
24 Deep Well Plate\n\nWell Shape: Square Top, V-Bottom\nVolume: 15mL Per Well\nMaterial: Polypropylene\nDimensions (mm): 127.2L  x  85.3W  x  63.2H\n\nQty/Pkg: 50
Catalog No. 50-008-04610 Supplier Analytical Sales & Services Supplier No. 24128
Add to Cart
Add to Cart
24 Deep Well Plate\n\nWell Shape: Square Top, V-Bottom\nVolume: 15mL Per Well\nMaterial: Polypropylene\nDimensions (mm): 127.2L  x  85.3W  x  63.2H\n\nQty/Pkg: 50